Tisk…

4-Chloro-5-methyl-5,6-dihydropyrido[2,3-d]pyrimidin-7(8H)-one

Dodavatel: Apollo Scientific

OR471461-1G OR471461-250MG
APOSOR471461-1GEA 31910 CZK
APOSOR471461-1G APOSOR471461-250MG
4-Chloro-5-methyl-5,6-dihydropyrido[2,3-d]pyrimidin-7(8H)-one
4-Chloro-5-methyl-5,6-dihydropyrido[2,3-d]pyrimidin-7(8H)-one
Vzorec: C₈H₈ClN₃O
Storage Temperature: Ambient
MDL Number: MFCD23098931
CAS číslo: 1177447-31-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR