2-(Hydroxymethyl)-5-methoxypyrimidine
Dodavatel: Apollo Scientific
OR471459-1G
OR471459-250MG
OR471459-5G
APOSOR471459-1GEA
16900
CZK
APOSOR471459-1G
APOSOR471459-250MG
APOSOR471459-5G
2-(Hydroxymethyl)-5-methoxypyrimidine
2-(Hydroxymethyl)-5-methoxypyrimidine
Vzorec:
C₆H₈N₂O₂ MW: 140.14 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD28501385 CAS číslo: 1459748-95-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...