Tisk…

2-(Boc-aminomethyl)benzyl Bromide

Dodavatel: Apollo Scientific

OR471454-1G OR471454-250MG OR471454-5G
APOSOR471454-1GEA 17130 CZK
APOSOR471454-1G APOSOR471454-250MG APOSOR471454-5G
2-(Boc-aminomethyl)benzyl Bromide
2-(Boc-aminomethyl)benzyl Bromide
Vzorec: C₁₃H₁₈BrNO₂
Storage Temperature: Ambient
MDL Number: MFCD20257492
CAS číslo: 220364-33-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR