2-Amino-4'-methoxybenzophenone
Dodavatel: Apollo Scientific
OR471451-1G
OR471451-250MG
OR471451-500MG
APOSOR471451-1GEA
27250
CZK
APOSOR471451-1G
APOSOR471451-250MG
APOSOR471451-500MG
2-Amino-4'-methoxybenzophenone
2-Amino-4'-methoxybenzophenone
Vzorec:
C₁₄H₁₃NO₂ MW: 227,259 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00034063 CAS číslo: 36192-61-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...