Tisk…

3-(4-Chlorophenyl)-2-hydroxypropionic acid

Dodavatel: Apollo Scientific
Varování

OR471447-5G OR471447-1G
APOSOR471447-5GEA 34390 CZK
APOSOR471447-5G APOSOR471447-1G
3-(4-Chlorophenyl)-2-hydroxypropionic acid
3-(4-Chlorophenyl)-2-hydroxypropionic acid
Vzorec: C₉H₉ClO₃
Storage Temperature: Ambient
MDL Number: MFCD11935789
CAS číslo: 23434-95-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR