(S)-4-Isobutyloxazolidin-2-one
Dodavatel: Apollo Scientific
OR471431-5G
OR471431-1G
APOSOR471431-5GEA
26090
CZK
APOSOR471431-5G
APOSOR471431-1G
(S)-4-Isobutyloxazolidin-2-one
(S)-4-Isobutyloxazolidin-2-one
Vzorec:
C₇H₁₃NO₂ Storage Temperature: Ambient |
MDL Number:
MFCD06656594 CAS číslo: 17016-85-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...