6-Hydrazinopyridazine-3-carboxamide
Dodavatel: Apollo Scientific
OR471388-1G
OR471388-250MG
APOSOR471388-1GEA
22920
CZK
APOSOR471388-1G
APOSOR471388-250MG
6-Hydrazinopyridazine-3-carboxamide
6-Hydrazinopyridazine-3-carboxamide
Vzorec:
C₅H₇N₅O MW: 153,142 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD16698821 CAS číslo: 3614-47-9 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...