Tisk…

(1S,2R)-2-Methylcyclohexylamine hydrochloride

Dodavatel: Apollo Scientific

Synonyms: (1S,2R)-2-Methylcyclohexanamine hydrochloride

OR471351-1G OR471351-250MG
APOSOR471351-1GEA 9230 CZK
APOSOR471351-1G APOSOR471351-250MG
(1S,2R)-2-Methylcyclohexylamine hydrochloride
(1S,2R)-2-Methylcyclohexylamine hydrochloride
Vzorec: C₇H₁₅N·ClH
MW: 149,662 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22201093
CAS číslo: 79389-39-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR