Tisk…

2-Acetyl-3-(Boc-amino)pyrazine

Dodavatel: Apollo Scientific

Synonyms: tert-Butyl (3-acetylpyrazin-2-yl)carbamate

OR471345-1G OR471345-250MG
APOSOR471345-1GEA 8670 CZK
APOSOR471345-1G APOSOR471345-250MG
2-Acetyl-3-(Boc-amino)pyrazine
2-Acetyl-3-(Boc-amino)pyrazine
Vzorec: C₁₁H₁₅N₃O₃
Storage Temperature: Ambient
MDL Number: MFCD28963469
CAS číslo: 1799412-29-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR