Tisk…

N-Boc-3-chloro-2-nitroaniline

Dodavatel: Apollo Scientific

OR471344-1G OR471344-250MG
APOSOR471344-1GEA 22920 CZK
APOSOR471344-1G APOSOR471344-250MG
N-Boc-3-chloro-2-nitroaniline
N-Boc-3-chloro-2-nitroaniline
Vzorec: C₁₁H₁₃ClN₂O₄
MW: 272,685 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19689610
CAS číslo: 1283176-45-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR