Tisk…

2-Chloro-4-nitro-m-phenylenediamine

Dodavatel: Apollo Scientific

OR471343-1G OR471343-5G
APOSOR471343-1GEA 11310 CZK
APOSOR471343-1G APOSOR471343-5G
2-Chloro-4-nitro-m-phenylenediamine
2-Chloro-4-nitro-m-phenylenediamine
Vzorec: C₆H₆ClN₃O₂
MW: 187,584 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08460155
CAS číslo: 261764-92-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR