5H-Indeno[1,2-c]pyridin-5-one
Dodavatel: Apollo Scientific
OR471340-1G
OR471340-250MG
APOSOR471340-1GEA
28700
CZK
APOSOR471340-1G
APOSOR471340-250MG
5H-Indeno[1,2-c]pyridin-5-one
5H-Indeno[1,2-c]pyridin-5-one
Vzorec:
C₁₂H₇NO MW: 181,19 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00757012 CAS číslo: 18631-22-6 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...