4-(2-Bromophenyl)-N,N-dimethylimidazole-1-sulphonamide
Dodavatel: Apollo Scientific
OR471338-1G
OR471338-250MG
APOSOR471338-1GEA
26090
CZK
APOSOR471338-1G
APOSOR471338-250MG
4-(2-Bromophenyl)-N,N-dimethylimidazole-1-sulphonamide
4-(2-Bromophenyl)-N,N-dimethylimidazole-1-sulphonamide
Vzorec:
C₁₁H₁₂BrN₃O₂S MW: 330,201 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD28968739 CAS číslo: 2006277-15-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...