Tisk…

Methyl 2-(Boc-amino)-2-(2-pyridyl)acetate

Dodavatel: Apollo Scientific

OR471330-1G OR471330-250MG
APOSOR471330-1GEA 20300 CZK
APOSOR471330-1G APOSOR471330-250MG
Methyl 2-(Boc-amino)-2-(2-pyridyl)acetate
Methyl 2-(Boc-amino)-2-(2-pyridyl)acetate
Vzorec: C₁₃H₁₈N₂O₄
MW: 266,293 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22576929
CAS číslo: 1001426-31-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR