Tisk…

4-Chloro-5-methoxybenzimidazole

Dodavatel: Apollo Scientific

OR471329-1G OR471329-250MG OR471329-5G
APOSOR471329-1GEA 20300 CZK
APOSOR471329-1G APOSOR471329-250MG APOSOR471329-5G
4-Chloro-5-methoxybenzimidazole
4-Chloro-5-methoxybenzimidazole
Vzorec: C₈H₇ClN₂O
MW: 182,607 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22558062
CAS číslo: 1360953-02-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR