1-Benzofuran-3-carbonitrile
Dodavatel: Apollo Scientific
OR471326-1G
OR471326-250MG
APOSOR471326-1GEA
26090
CZK
APOSOR471326-1G
APOSOR471326-250MG
1-Benzofuran-3-carbonitrile
1-Benzofuran-3-carbonitrile
Vzorec:
C₉H₅NO MW: 143,142 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00810098 CAS číslo: 55877-31-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...