Tisk…

7-(Aminomethyl)benzoxazole hydrochloride

Dodavatel: Apollo Scientific

OR471321-1G OR471321-250MG
APOSOR471321-1GEA 40670 CZK
APOSOR471321-1G APOSOR471321-250MG
7-(Aminomethyl)benzoxazole hydrochloride
7-(Aminomethyl)benzoxazole hydrochloride
Vzorec: C₈H₈N₂O·ClH
MW: 184,623 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28954615
CAS číslo: 2006276-90-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR