(R)-2-Phenylpropanal
Dodavatel: Apollo Scientific
OR471318-1G
OR471318-250MG
APOSOR471318-1GEA
17690
CZK
APOSOR471318-1G
APOSOR471318-250MG
(R)-2-Phenylpropanal
(R)-2-Phenylpropanal
Vzorec:
C₉H₁₀O Storage Temperature: Ambient |
MDL Number:
MFCD24673693 CAS číslo: 38235-74-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...