Tisk…

2-Methylquinoline-5-boronic acid pinacol ester

Dodavatel: Apollo Scientific

Synonyms: 2-Methylquinoline-5-boronic Acid Pinacol Ester

OR471311-1G OR471311-250MG
APOSOR471311-1GEA 10520 CZK
APOSOR471311-1G APOSOR471311-250MG
2-Methylquinoline-5-boronic acid pinacol ester
2-Methylquinoline-5-boronic acid pinacol ester
Vzorec: C₁₆H₂₀BNO₂
MW: 269.146 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22418301
CAS číslo: 876922-75-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR