Tisk…

3-(Aminomethyl)-2-methylpyridine

Dodavatel: Apollo Scientific

OR471300-5G OR471300-1G
APOSOR471300-5GEA 69760 CZK
APOSOR471300-5G APOSOR471300-1G
3-(Aminomethyl)-2-methylpyridine
3-(Aminomethyl)-2-methylpyridine
Vzorec: C₇H₁₀N₂
MW: 122.168 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03411326
CAS číslo: 58539-64-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR