Tisk…

3-(Aminomethyl)-5-bromo-2-methylpyridine

Dodavatel: Apollo Scientific

OR471293-1G OR471293-250MG
APOSOR471293-1GEA 26090 CZK
APOSOR471293-1G APOSOR471293-250MG
3-(Aminomethyl)-5-bromo-2-methylpyridine
3-(Aminomethyl)-5-bromo-2-methylpyridine
Vzorec: C₇H₉BrN₂
MW: 201,064 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18255107
CAS číslo: 1211589-61-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR