4-(Aminomethyl)-5-chloro-2-(methylthio)pyrimidine
Dodavatel: Apollo Scientific
OR471259-1G
OR471259-250MG
APOSOR471259-1GEA
31910
CZK
APOSOR471259-1G
APOSOR471259-250MG
4-(Aminomethyl)-5-chloro-2-(methylthio)pyrimidine
4-(Aminomethyl)-5-chloro-2-(methylthio)pyrimidine
Vzorec:
C₆H₈ClN₃S Storage Temperature: Ambient |
MDL Number:
MFCD28398152 CAS číslo: 1511510-10-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...