2,5-Dichloroquinoline-3-carbaldehyde
Dodavatel: Apollo Scientific
OR471195-1G
OR471195-250MG
APOSOR471195-1GEA
22920
CZK
APOSOR471195-1G
APOSOR471195-250MG
2,5-Dichloroquinoline-3-carbaldehyde
2,5-Dichloroquinoline-3-carbaldehyde
Vzorec:
C₁₀H₅Cl₂NO Storage Temperature: Ambient |
MDL Number:
MFCD22370818 CAS číslo: 1064137-50-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...