Tisk…

4-(3-Bromopropyl)morpholine hydrobromide

Dodavatel: Apollo Scientific

OR471189-1G OR471189-5G
APOSOR471189-1GEA 3110 CZK
APOSOR471189-1G APOSOR471189-5G
4-(3-Bromopropyl)morpholine hydrobromide
4-(3-Bromopropyl)morpholine hydrobromide
Vzorec: C₇H₁₄BrNO·BrH
Storage Temperature: Ambient
MDL Number: MFCD13176498
CAS číslo: 88806-06-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR