Methyl 2-(2-Cyano-4-hydroxyphenyl)acetate
Dodavatel: Apollo Scientific
OR471181-1G
OR471181-250MG
APOSOR471181-1GEA
43640
CZK
APOSOR471181-1G
APOSOR471181-250MG
Methyl 2-(2-Cyano-4-hydroxyphenyl)acetate
Methyl 2-(2-Cyano-4-hydroxyphenyl)acetate
Vzorec:
C₁₀H₉NO₃ MW: 191.183 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD18399075 CAS číslo: 1261559-90-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...