Tisk…

Methyl 2-(2-Bromo-4-pyridyl)acetate

Dodavatel: Apollo Scientific

OR471146-1G OR471146-5G
APOSOR471146-1GEA 7280 CZK
APOSOR471146-1G APOSOR471146-5G
Methyl 2-(2-Bromo-4-pyridyl)acetate
Methyl 2-(2-Bromo-4-pyridyl)acetate
Vzorec: C₈H₈BrNO₂
MW: 230,059 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18255863
CAS číslo: 1234217-58-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR