Tisk…

2-(Boc-amino)-2-phenylethylamine hydrochloride

Dodavatel: Apollo Scientific

OR471115-5G OR471115-1G
APOSOR471115-5GEA 24370 CZK
APOSOR471115-5G APOSOR471115-1G
2-(Boc-amino)-2-phenylethylamine hydrochloride
2-(Boc-amino)-2-phenylethylamine hydrochloride
Vzorec: C₁₃H₂₀N₂O₂·ClH
MW: 272,771 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28144002
CAS číslo: 1253955-71-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR