Tisk…

2-Amino-6-chloroquinoline

Dodavatel: Apollo Scientific

OR471112-1G OR471112-250MG
APOSOR471112-1GEA 22920 CZK
APOSOR471112-1G APOSOR471112-250MG
2-Amino-6-chloroquinoline
2-Amino-6-chloroquinoline
Vzorec: C₉H₇ClN₂
Storage Temperature: Ambient
MDL Number: MFCD08235108
CAS číslo: 18672-02-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR