Methyl 4-bromo-2,6-dinitrobenzoate
Dodavatel: Apollo Scientific
OR471103-1G
OR471103-5G
OR471103-250MG
APOSOR471103-1GEA
12770
CZK
APOSOR471103-1G
APOSOR471103-5G
APOSOR471103-250MG
Methyl 4-bromo-2,6-dinitrobenzoate
Methyl 4-bromo-2,6-dinitrobenzoate
Vzorec:
C₈H₅BrN₂O₆ MW: 305.039 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD18651710 CAS číslo: 1272756-03-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...