Ethyl 3-Amino-4-hydroxybenzoate
Dodavatel: Apollo Scientific
|
Varování
|
OR471083-5G
OR471083-1G
OR471083-10G
APOSOR471083-5GEA
8040
CZK
APOSOR471083-5G
APOSOR471083-1G
APOSOR471083-10G
Ethyl 3-Amino-4-hydroxybenzoate
Ethyl 3-Amino-4-hydroxybenzoate
Vzorec:
C₉H₁₁NO₃ MW: 181,189 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09961411 CAS číslo: 13052-92-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...