Tisk…

2-(2-Acetyl-4-bromophenoxy)acetic acid

Dodavatel: Apollo Scientific

OR471080-5G OR471080-1G
APOSOR471080-5GEA 28110 CZK
APOSOR471080-5G APOSOR471080-1G
2-(2-Acetyl-4-bromophenoxy)acetic acid
2-(2-Acetyl-4-bromophenoxy)acetic acid
Vzorec: C₁₀H₉BrO₄
MW: 273.08 g/mol
Storage Temperature: Ambient
MDL Number: MFCD27949767
CAS číslo: 34849-51-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR