Tisk…

2-Bromo-4-methoxybenzyl Alcohol

Dodavatel: Apollo Scientific
Varování

Synonyms: (2-Bromo-4-methoxy-phenyl)methanol

OR471018-5G OR471018-1G
APOSOR471018-5GEA 10820 CZK
APOSOR471018-5G APOSOR471018-1G
2-Bromo-4-methoxybenzyl Alcohol
2-Bromo-4-methoxybenzyl Alcohol
Vzorec: C₈H₉BrO₂
MW: 217.06 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11042785
CAS číslo: 163190-79-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR