3-Ethoxy-5-methylbenzoic acid
Dodavatel: Apollo Scientific
OR471007-1G
OR471007-250MG
OR471007-5G
APOSOR471007-1GEA
13060
CZK
APOSOR471007-1G
APOSOR471007-250MG
APOSOR471007-5G
3-Ethoxy-5-methylbenzoic acid
3-Ethoxy-5-methylbenzoic acid
Vzorec:
C₁₀H₁₂O₃ MW: 180,2 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD27940681 CAS číslo: 500901-64-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...