Tisk…

2-Chloro-5-(1-piperazinyl)pyrazine

Dodavatel: Apollo Scientific

OR470999-1G OR470999-250MG OR470999-5G
APOSOR470999-1GEA 26090 CZK
APOSOR470999-1G APOSOR470999-250MG APOSOR470999-5G
2-Chloro-5-(1-piperazinyl)pyrazine
2-Chloro-5-(1-piperazinyl)pyrazine
Vzorec: C₈H₁₁ClN₄
MW: 198,653 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05182228
CAS číslo: 84445-51-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR