Tisk…

2-Chloro-4-isopropoxybenzoic acid

Dodavatel: Apollo Scientific

Synonyms: 2-Chloro-4-isopropoxybenzoic Acid

OR470995-1G OR470995-5G
APOSOR470995-1GEA 7280 CZK
APOSOR470995-1G APOSOR470995-5G
2-Chloro-4-isopropoxybenzoic acid
2-Chloro-4-isopropoxybenzoic acid
Vzorec: C₁₀H₁₁ClO₃
MW: 214,646 g/mol
Storage Temperature: Ambient
MDL Number: MFCD17256599
CAS číslo: 334018-32-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR