Tisk…

3-Chloro-4-(cyclopropylmethoxy)benzoic acid

Dodavatel: Apollo Scientific

OR470994-1G OR470994-5G
APOSOR470994-1GEA 8700 CZK
APOSOR470994-1G APOSOR470994-5G
3-Chloro-4-(cyclopropylmethoxy)benzoic acid
3-Chloro-4-(cyclopropylmethoxy)benzoic acid
Vzorec: C₁₁H₁₁ClO₃
MW: 226,656 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14652993
CAS číslo: 856165-89-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR