Tisk…

Methyl 3-Bromobenzimidate hydrochloride

Dodavatel: Apollo Scientific

OR470983-1G OR470983-5G
APOSOR470983-1GEA 7280 CZK
APOSOR470983-1G APOSOR470983-5G
Methyl 3-Bromobenzimidate hydrochloride
Methyl 3-Bromobenzimidate hydrochloride
Vzorec: C₈H₈BrNO·ClH
MW: 250,52 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10697946
CAS číslo: 56108-13-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR