Tisk…

6-Amino-9-(tetrahydropyran-2-yl)-9H-purine

Dodavatel: Apollo Scientific

Synonyms: 9-(Tetrahydro-2H-pyran-2-yl)-9H-purin-6-amine

OR470947-1G OR470947-250MG OR470947-5G
APOSOR470947-1GEA 3970 CZK
APOSOR470947-1G APOSOR470947-250MG APOSOR470947-5G
6-Amino-9-(tetrahydropyran-2-yl)-9H-purine
6-Amino-9-(tetrahydropyran-2-yl)-9H-purine
Vzorec: C₁₀H₁₃N₅O
MW: 219,243 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00213333
CAS číslo: 7306-67-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR