Tisk…

Ethyl 6-Bromoisoquinoline-1-carboxylate

Dodavatel: Apollo Scientific
Varování

OR470934-1G OR470934-250MG
APOSOR470934-1GEA 12340 CZK
APOSOR470934-1G APOSOR470934-250MG
Ethyl 6-Bromoisoquinoline-1-carboxylate
Ethyl 6-Bromoisoquinoline-1-carboxylate
Vzorec: C₁₂H₁₀BrNO₂
MW: 280,117 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18257309
CAS číslo: 1020576-70-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR