Tisk…

2-Cyclopropylpyridine-3-boronic acid pinacol ester

Dodavatel: Apollo Scientific

OR470925-1G OR470925-250MG
APOSOR470925-1GEA 26090 CZK
APOSOR470925-1G APOSOR470925-250MG
2-Cyclopropylpyridine-3-boronic acid pinacol ester
2-Cyclopropylpyridine-3-boronic acid pinacol ester
Vzorec: C₁₄H₂₀BNO₂
MW: 245,125 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18733626
CAS číslo: 2098215-65-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR