5-Hydroxy-6-oxo-1,6-dihydropyridine-3-carboxylic acid 97%
Dodavatel: Apollo Scientific
OR470875-1G
OR470875-250MG
APOSOR470875-1GEA
26920
CZK
APOSOR470875-1G
APOSOR470875-250MG
5-Hydroxy-6-oxo-1,6-dihydropyridine-3-carboxylic acid 97%
5-Hydroxy-6-oxo-1,6-dihydropyridine-3-carboxylic acid
Vzorec:
C₆H₅NO₄ MW: 155,108 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD16659014 CAS číslo: 76470-35-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...