2-(Benzylamino)-3-hydroxy-3-methylbutanoic acid
Dodavatel: Apollo Scientific
OR470843-5G
OR470843-1G
APOSOR470843-5GEA
22920
CZK
APOSOR470843-5G
APOSOR470843-1G
2-(Benzylamino)-3-hydroxy-3-methylbutanoic acid
2-(Benzylamino)-3-hydroxy-3-methylbutanoic acid
Vzorec:
C₁₂H₁₇NO₃ MW: 223,268 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD27935371 CAS číslo: 14598-96-0 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...