Methyl 2-(1-Boc-4-piperidylidene)-2-(Cbz-amino)acetate
Dodavatel: Apollo Scientific
OR470833-1G
OR470833-5G
APOSOR470833-1GEA
13060
CZK
APOSOR470833-1G
APOSOR470833-5G
Methyl 2-(1-Boc-4-piperidylidene)-2-(Cbz-amino)acetate
Methyl 2-(1-Boc-4-piperidylidene)-2-(Cbz-amino)acetate
Vzorec:
C₂₁H₂₈N₂O₆ MW: 404,457 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD23702003 CAS číslo: 313491-20-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...