Tisk…

N-Ethylcyclopropanamine hydrochloride

Dodavatel: Apollo Scientific
Varování

OR470784-1G OR470784-5G
APOSOR470784-1GEA 9590 CZK
APOSOR470784-1G APOSOR470784-5G
N-Ethylcyclopropanamine hydrochloride
N-Ethylcyclopropanamine hydrochloride
Vzorec: C₅H₁₁N·ClH
Storage Temperature: Ambient
MDL Number: MFCD11099458
CAS číslo: 672302-35-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR