Tisk…

5-Bromo-7-methoxybenzofuran

Dodavatel: Apollo Scientific

OR470779-1G OR470779-250MG OR470779-5G
APOSOR470779-1GEA 22920 CZK
APOSOR470779-1G APOSOR470779-250MG APOSOR470779-5G
5-Bromo-7-methoxybenzofuran
5-Bromo-7-methoxybenzofuran
Vzorec: C₉H₇BrO₂
MW: 227,055 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11877831
CAS číslo: 164414-62-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR