Tisk…

3-(Chloromethyl)-1-methylpyrrolidine hydrochloride

Dodavatel: Apollo Scientific

OR470762-1G OR470762-5G
APOSOR470762-1GEA 18820 CZK
APOSOR470762-1G APOSOR470762-5G
3-(Chloromethyl)-1-methylpyrrolidine hydrochloride
3-(Chloromethyl)-1-methylpyrrolidine hydrochloride
Vzorec: C₆H₁₂ClN·HCl
MW: 170,08 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20441425
CAS číslo: 56669-84-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR