Tisk…

2-(Aminomethyl)benzoxazole

Dodavatel: Apollo Scientific

OR470756-250MG OR470756-1G
APOSOR470756-250MGEA 8700 CZK
APOSOR470756-250MG APOSOR470756-1G
2-(Aminomethyl)benzoxazole
2-(Aminomethyl)benzoxazole
Vzorec: C₈H₈N₂O
MW: 148,162 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03990563
CAS číslo: 101333-98-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR