Methyl (S)-2-(Boc-amino)-3-(1-methyl-5-imidazolyl)propanoate
Dodavatel: Apollo Scientific
OR470741-1G
OR470741-250MG
OR470741-5G
APOSOR470741-1GEA
28700
CZK
APOSOR470741-1G
APOSOR470741-250MG
APOSOR470741-5G
Methyl (S)-2-(Boc-amino)-3-(1-methyl-5-imidazolyl)propanoate
Methyl (S)-2-(Boc-amino)-3-(1-methyl-5-imidazolyl)propanoate
Vzorec:
C₁₃H₂₁N₃O₄ MW: 283,324 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD16250467 CAS číslo: 72212-51-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...