Tisk…

2-Cyano-3-(4-hydroxyphenyl)propanoic acid

Dodavatel: Apollo Scientific

OR470737-5G OR470737-1G
APOSOR470737-5GEA 37690 CZK
APOSOR470737-5G APOSOR470737-1G
2-Cyano-3-(4-hydroxyphenyl)propanoic acid
2-Cyano-3-(4-hydroxyphenyl)propanoic acid
Vzorec: C₁₀H₉NO₃
MW: 191,183 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11593027
CAS číslo: 90924-41-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR