3-(4-Hydroxy-3,5-dimethoxyphenyl)propanoic acid
Dodavatel: Apollo Scientific
OR470736-5G
OR470736-1G
APOSOR470736-5GEA
31910
CZK
APOSOR470736-5G
APOSOR470736-1G
3-(4-Hydroxy-3,5-dimethoxyphenyl)propanoic acid
3-(4-Hydroxy-3,5-dimethoxyphenyl)propanoic acid
Vzorec:
C₁₁H₁₄O₅ MW: 226,226 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00016559 CAS číslo: 14897-78-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...